missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | PSMD6 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230879
|
Novus Biologicals
NBP3-35811-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229750
|
Novus Biologicals
NBP3-35811-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSMD6 Polyclonal antibody specifically detects PSMD6 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| PSMD6 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| 26S proteasome non-ATPase regulatory subunit 6, Breast cancer-associated protein SGA-113M, KIAA010726S proteasome regulatory subunit S10, p42A, p44S10, PFAAP4, Phosphonoformate immuno-associated protein 4, proteasome (prosome, macropain) 26S subunit, non-ATPase, 6,26S proteasome regulatory subunit RPN7, Proteasome regulatory particle subunit p44S10, Rpn7, S10, SGA-113M | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 23-307 of human PSMD6 (NP_055629.1).,, Sequence:, RFLLSLPEHRGDAAVRDELMAAVRDNNMAPYYEALCKSLDWQIDVDLLNKMKKANEDELKRLDEELEDAEKNLGESEIRDAMMAKAEYLCRIGDKEGALTAFRKTYDKTVALGHRLDIVFYLLRIGLFYMDNDLITRNTEKAKSLIEEGGDWDRRNRLKVYQGLYCVAIRDFKQAAELFLDTVSTFTSYELMDYKTFVTYTVYVSMIALERPDLREKVIKGAEILEVLHSLPAVRQYLFSLYECRYSVFFQSLAVVEQEMKKDWLFAPHYRYYVREMRIHAYSQL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 9861 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title