missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PSMD14 Polyclonal antibody specifically detects PSMD14 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PSMD14. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?