missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSMD12 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15742631 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15742631 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15742631 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' PA554701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: WQQALKSVVL YVILAPFDNE QSDLVHRISG DKKLEEIPKY KDLLKLFTTM ELMRWSTLVE DYGMELRKGS LESPATDVFG STEEGEKRWK DLKNRVVEH Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

This protein is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that binds directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein is found in many histone deacetylase complexes, including mSin3 co-repressor complex. It is also present in protein complexes involved in chromatin assembly. This protein can interact with BRCA1 tumor-suppressor gene and may have a role in the regulation of cell proliferation and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMD12
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSMD12
Gene Accession No. O00232
Gene Alias 1500002F15Rik; 26S proteasome non-ATPase regulatory subunit 12; 26S proteasome non-ATPase regulatory subunit 1-like protein; 26S proteasome regulatory subunit p55; 26S proteasome regulatory subunit RPN5; AI480719; fc34f08; MGC75406; P55; proteasome (prosome, macropain) 26S subunit, non-ATPase, 12; proteasome 26S non-ATPase subunit 12; proteasome 26S subunit non-ATPase 12; proteasome 26S subunit, non-ATPase 12; PSMD12; psmd12 protein; Rpn5; similar to proteasome 26S non-ATPase subunit 12 isoform 2; wu:fb38a06; wu:fc34f08; wu:fc49f02
Gene Symbols PSMD12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSMD12. Recombinant protein control fragment (Product #RP-93548).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5718
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.