missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PSMD12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29923
This item is not returnable.
View return policy
Description
PSMD12 Polyclonal antibody specifically detects PSMD12 in Human samples. It is validated for Immunoprecipitation
Specifications
| PSMD12 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| 26S proteasome non-ATPase regulatory subunit 12, 26S proteasome regulatory subunit p55, 26S proteasome regulatory subunit RPN5, MGC75406, p55, proteasome (prosome, macropain) 26S subunit, non-ATPase, 12, Rpn5 | |
| Between 406 and 456 | |
| 100 μg | |
| Cell Biology | |
| 5718 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction