missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMD11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550566
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PSMD11 protein.
The 26S proteasome (PSMD11) is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. The protein is a non-ATPase subunit of the 19S regulator.
Specifications
| PSMD11 | |
| Polyclonal | |
| Unconjugated | |
| Psmd11 | |
| 1700089D09Rik; 1810019E17Rik; 2610024G20Rik; 26S proteasome non-ATPase regulatory subunit 11; 26S proteasome regulatory subunit 9; 26S proteasome regulatory subunit p44.5; 26S proteasome regulatory subunit RPN6; 26S proteasome regulatory subunit S9; 2810055C24Rik; C78232; p44.5; proteasome (prosome, macropain) 26S subunit, non-ATPase, 11; proteasome 26S subunit, non-ATPase 11; Psmd11; Rpn6; S9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 303353, 5717, 69077 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 2.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| F1LMZ8, O00231, Q8BG32 | |
| Psmd11 | |
| Synthetic peptide corresponding to a region derived from internal residues of human proteasome (prosome, macropain) 26S subunit, non-ATPase, 11. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction