missing translation for 'onlineSavingsMsg'
Learn More

PSMD11 Antibody (2C7), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Bio-Techne NBP2-59484AF750

Product Code. 18249530

  • £442.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PSMD11 Monoclonal antibody specifically detects PSMD11 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PSMD11
Monoclonal
Alexa Fluor 750
MGC3844,26S proteasome regulatory subunit RPN6, p44.5,26S proteasome regulatory subunit 9, proteasome (prosome, macropain) 26S subunit, non-ATPase, 11,26S proteasome regulatory subunit p44.5, Rpn6,26S proteasome regulatory subunit S9, S9,26S proteasome non-ATPase regulatory subunit 11
Recombinant human PSMD11 (1-422aa) purified from E. coli
0.1 mL
Primary
Human
Purified
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
2C7
50mM Sodium Borate
Mouse
Protein A purified
RUO
5717
Store at 4°C in the dark.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.