missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMC6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102754
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PSMC6.
PSMC6 (proteasome (prosome, macropain) 26S subunit, ATPase 6), also known as P44, p42, SUG2, S10B or CADP44, is a regulatory component of the 26S proteasome. More specifically, PSMC6 is one of the six ATPase subunits of the 19S regulator base. It contains a leucine zipper motif and an AAA (ATPase associated with diverse cellular activities) domain and belongs to the AAA ATPase family of chaperone-like ATPases.
Specifications
| PSMC6 | |
| Polyclonal | |
| Unconjugated | |
| PSMC6 | |
| 2300001E01Rik; 26S protease regulatory subunit 10B; 26S protease regulatory subunit S10B; 26S proteasome AAA-ATPase subunit RPT4; 26S proteasome regulatory subunit 10B; AI451058; CADP44; conserved ATPase domain protein 44; epididymis secretory protein Li 73; HEL-S-73; p42; P44; proteasome (prosome, macropain) 26S subunit, ATPase, 6; proteasome (prosome, macropain) 26S subunit, ATPase, 6-like; proteasome 26S subunit ATPase 6; proteasome 26S subunit, ATPase 6; proteasome subunit p42; PSMC6; SUG2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 289990, 5706, 67089 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62333, P62334 | |
| PSMC6 | |
| A synthesized peptide derived from human PSMC6(Accession P62333), corresponding to amino acid residues F219-T269. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction