missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99613-50ul
This item is not returnable.
View return policy
Description
PSMC6 Polyclonal antibody specifically detects PSMC6 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PSMC6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:200-1:1000 | |
| 26S protease regulatory subunit 10B, 26S protease regulatory subunit S10B, 26S proteasome AAA-ATPase subunit RPT4, CADP44, conserved ATPase domain protein 44, MGC12520, p42, P44, proteasome (prosome, macropain) 26S subunit, ATPase, 6, Proteasome 26S subunit ATPase 6, Proteasome subunit p42, SUG2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PSMC6. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 5706 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction