missing translation for 'onlineSavingsMsg'
Learn More

PSMC6 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18370314 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370314 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370314 Supplier Novus Biologicals Supplier No. NBP299613IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSMC6 Polyclonal specifically detects PSMC6 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMC6
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias 26S protease regulatory subunit 10B, 26S protease regulatory subunit S10B, 26S proteasome AAA-ATPase subunit RPT4, CADP44, conserved ATPase domain protein 44, MGC12520, p42, P44, proteasome (prosome, macropain) 26S subunit, ATPase, 6, Proteasome 26S subunit ATPase 6, Proteasome subunit p42, SUG2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human PSMC6.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5706
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.