Learn More
Invitrogen™ PSMC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530133
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (100%), Pig (100%), Chicken (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Human SUG1 (also known as p45, Rpt6 and Cim3) is a component of the 19S regulatory subunit of the 26S proteosome complex. It is one of the AAA ATPas found in the 19S regulatory complex that is thought to participate in the unfolding of ubiquitinated proteins in an ATP-dependent manner. It has also been shown to directly interact with the nucleotide excision repair protein XPB.
Specifications
| PSMC5 | |
| Polyclonal | |
| Unconjugated | |
| Psmc5 | |
| 26S protease regulatory subunit 8; 26S proteasome AAA-ATPase subunit RPT6; 26S proteasome regulatory subunit 8; for proteasomal ATPase (SUG1); mSUG1; MSUG1 protein; p45; p45/SUG; peptidase (prosome, macropain) 26S subunit, ATPase 5; protease (prosome, macropain) 26S subunit, ATPase 5; proteasomal ATPase (SUG1); proteasome (prosome, macropain) 26S subunit, ATPase, 5; proteasome 26S ATPase subunit 5; proteasome 26S subunit ATPase 5; proteasome 26S subunit, ATPase 5; proteasome p45/SUG; proteasome subunit p45; Psmc5; S8; SUG1; SUG-1; Tat-binding protein homolog 10; TBP10; testicular tissue protein Li 149; thy; thyroid hormone receptor-interacting protein 1; thyroid receptor interactor 1; TRIP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19184, 5705 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.73 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P62195, P62196 | |
| Psmc5 | |
| Recombinant protein encompassing a sequence within the center region of human PSMC5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.