missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMC3IP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100560
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HOP2.
PSMC3IP plays an important role in meiotic recombination. It stimulates DMC1-mediated strand exchange required for pairing homologous chromosomes during meiosis. The complex PSMC3IP/MND1 binds DNA, stimulates the recombinase activity of DMC1 as well as DMC1 D-loop formation from double-strand DNA. This complex stabilizes presynaptic RAD51 and DMC1 filaments formed on single strand DNA to capture double-strand DNA. This complex stimulates both synaptic and presynaptic critical steps in RAD51 and DMC1-promoted homologous pairing. It may inhibit HIV-1 viral protein TAT activity and modulate the activity of proteasomes through association with PSMC3.
Specifications
| PSMC3IP | |
| Polyclonal | |
| Unconjugated | |
| PSMC3IP | |
| C79099; DBD-interacting; Gt198; Homologous-pairing protein 2 homolog; homologous-pairing protein 2 homolog-like; Hop2; HUMGT198A; Nuclear receptor coactivator GT198; ODG3; proteasome (prosome, macropain) 26S subunit, ATPase 3, interacting protein; proteasome 26S ATPase subunit 3-interacting protein; PSMC3 interacting protein; PSMC3-interacting protein; PSMC3IP; Tat-binding protein 1-interacting protein; TBP-1-interacting protein; Tbpip | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29893 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9P2W1 | |
| PSMC3IP | |
| A synthesized peptide derived from human PSMC3IP(Accession Q9P2W1), corresponding to amino acid residues R166-D216. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction