missing translation for 'onlineSavingsMsg'
Learn More

PSMC3IP Antibody, Novus Biologicals™

Product Code. 18333689 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333689 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333689 Supplier Novus Biologicals Supplier No. H00029893B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

PSMC3IP Polyclonal antibody specifically detects PSMC3IP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMC3IP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_057640.1
Gene Alias DBD-interacting, GT198, GT198 alternative, homologous-pairing protein 2 homolog, HOP2, HUMGT198A, Nuclear receptor coactivator GT198, Proteasome 26S ATPase subunit 3-interacting protein, PSMC3 interacting protein, Tat-binding protein 1-interacting protein, TBP-1 interacting protein, TBP-1-interacting protein, TBPIPPSMC3-interacting protein
Host Species Mouse
Immunogen PSMC3IP (NP_057640.1, 1 a.a. - 217 a.a.) full-length human protein. MSKGRAEAAAGAAGILLRYLQEQNRPYSSQDVFGNLQREHGLGKAVVVKTLEQLAQQGKIKEKMYGKQKIYFADQDQFDMVSDADLQVLDGKIVALTAKVQSLQQSCRYMEAELKELSSALTTPEMQKEIQELKKECAGYRERLKNIKAATNHVTPEEKEQVYRERQKYCKEWRKRKRMATELSDAILEGYPKSKKQFFEEVGIETDEDYNVTLPDP
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29893
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.