missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC3 Mouse anti-Human, Monkey, Rat, Clone:1G10C9, Abnova™
Mouse monoclonal antibody raised against recombinant human PSMC3.
Brand: Abnova MAB17232.100ug
This item is not returnable.
View return policy
Description
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes one of the ATPase subunits, a member of the triple-A family of ATPases that have chaperone-like activity. This subunit may compete with PSMC2 for binding to the HIV tat protein to regulate the interaction between the viral protein and the transcription complex. A pseudogene has been identified on chromosome 9. [provided by RefSeq]Specifications
| PSMC3 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| PSMC3 | |
| Mouse | |
| 100 μg | |
| Human, Monkey, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 1G10C9 | |
| proteasome (prosome, macropain) 26S subunit, ATPase, 3 | |
| In PBS (0.05% sodium azide) | |
| MGC8487, TBP1 | |
| Recombinant protein corresponding to amino acid 53-152 of human PSMC3 from E. coli. | |
| 5702 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction