missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMC3 Monoclonal Antibody (OTI1C10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525047
This item is not returnable.
View return policy
Description
PSMC3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a WASP interacting protein (WIP)-related protein. It has been shown that this protein has a role in the WASP-mediated organization of the actin cytoskeleton and that this protein is a potential link between the activated platelet-derived growth factor receptor and the actin polymerization machinery.
Specifications
| PSMC3 | |
| Monoclonal | |
| 0.81 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P17980, Q63569 | |
| PSMC3 | |
| Full length human recombinant protein of PSMC3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Rat, Canine, Monkey | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1C10 | |
| Unconjugated | |
| PSMC3 | |
| 26S protease regulatory subunit 6A; 26S proteasome AAA-ATPase subunit RPT5; 26S proteasome regulatory subunit 6A; human immunodeficiency virus tat transactivator binding protein-1; MGC8487; proteasome (prosome, macropain) 26S subunit, ATPase 3; proteasome (prosome, macropain) 26S subunit, ATPase, 3; proteasome 26S subunit ATPase 3; proteasome 26S subunit, ATPase 3; proteasome subunit P50; Psmc3; rpt5; Spermatogenic cell/sperm-associated Tat-binding protein homolog SATA; Tat binding protein 1; Tat-binding protein 1; TBP1; TBP-1; testicular secretory protein Li 42 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29677, 475980, 5702 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction