missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35333-20ul
This item is not returnable.
View return policy
Description
PSMC3 Polyclonal antibody specifically detects PSMC3 in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| PSMC3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| human immunodeficiency virus tat transactivator binding protein-1, MGC8487, proteasome (prosome, macropain) 26S subunit, ATPase, 3,26S proteasome AAA-ATPase subunit RPT5, Proteasome 26S subunit ATPase 3, Proteasome subunit P50, TBP-126S protease regulatory subunit 6A, TBP1Tat-binding protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human PSMC3 (NP_002799.3).,, Sequence:, MEEGGRDKAPVQPQQSPAAAPGGTDEKPSGKERRDAGDKDKEQELSEEDKQLQDELEMLVERLGEKDTSLYRPALEELRRQIRSSTTSMTSVPKPLKFLRPHYGKLKEIYENMAPGENKRFAADIISVLAMTMSGERECLKYRLVGSQEELASWGHEYVRHLAGEVAKEWQELDDAEKVQ | |
| 20 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5702 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction