missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC3 Antibody (1G10B7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £453.00
Specifications
| Antigen | PSMC3 |
|---|---|
| Clone | 1G10B7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18247374
|
Novus Biologicals
NBP2-61802 |
0.1 mL |
£453.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604347
|
Novus Biologicals
NBP2-61802-0.025ml |
0.025 mL |
£173.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSMC3 Monoclonal specifically detects PSMC3 in Human, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PSMC3 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| human immunodeficiency virus tat transactivator binding protein-1, MGC8487, proteasome (prosome, macropain) 26S subunit, ATPase, 3,26S proteasome AAA-ATPase subunit RPT5, Proteasome 26S subunit ATPase 3, Proteasome subunit P50, TBP-126S protease regulatory subunit 6A, TBP1Tat-binding protein 1 | |
| PSMC3 | |
| IgG1 | |
| Protein G purified |
| 1G10B7 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| 5702 | |
| Purified recombinant fragment of human PSMC3 (AA: 53-152) expressed in E. Coli. | |
| Primary | |
| 49 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title