missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3317
This item is not returnable.
View return policy
Description
PSMC1 Polyclonal specifically detects PSMC1 in Human, Yeast samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| PSMC1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
| 26S protease regulatory subunit 4, 26S proteasome AAA-ATPase subunit RPT2, MGC24583, P26S4, p56, proteasome (prosome, macropain) 26S subunit, ATPase, 1, proteasome 26S ATPase subunit 1, Proteasome 26S subunit ATPase 1, proteasome 26S subunit, ATPase, 1, S4MGC8541 | |
| Rabbit | |
| 50 kDa | |
| 100 μL | |
| Primary | |
| Detects proteasome 19S subunit S4. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0mg/mL | |
| Western Blot 1:1000, Immunoprecipitation 1:10 - 1:500 | |
| P62191 | |
| PSMC1 | |
| Yeast proteasome 19S subunit S4. | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5700 | |
| Human, Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction