missing translation for 'onlineSavingsMsg'
Learn More

PSMC1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18420558 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420558 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420558 Supplier Novus Biologicals Supplier No. NBP214856AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSMC1 Polyclonal antibody specifically detects PSMC1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMC1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 26S protease regulatory subunit 4, 26S proteasome AAA-ATPase subunit RPT2, MGC24583, P26S4, p56, proteasome (prosome, macropain) 26S subunit, ATPase, 1, proteasome 26S ATPase subunit 1, Proteasome 26S subunit ATPase 1, proteasome 26S subunit, ATPase, 1, S4MGC8541
Host Species Rabbit
Immunogen This antibody maps to a region between residue 50 and 100 of human Proteasome (prosome, macropain) Subunit, ATPase, 1 using the numbering given in entry NP_002793.2 (GeneID 5700).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5700
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.