missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMB9 Polyclonal Antibody

Rabbit Polyclonal Antibody
£133.00 - £397.00
Specifications
| Antigen | PSMB9 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17740774
![]() |
Invitrogen™
PA5119368 |
200 μL |
£397.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17763234
|
Invitrogen™
PA5121838 |
20 μL |
£133.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSMB9 Polyclonal Antibody for Western Blot, IHC (P)
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. This gene is located in the class II region of the MHC in the immunoproteasome. Proteolytic processing is required to generate a mature subunit.Specifications
| PSMB9 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Psmb9 | |
| beta1i; large multifunctional peptidase 2; large multifunctional protease 2; LMP2; Lmp-2; LMP-2d; low molecular mass polypeptide 2; low molecular mass protein 2; Macropain chain 7; MGC70470; multicatalytic endopeptidase complex chain 7; proteasome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2); proteasome (prosome, macropain) subunit, beta type, 9; proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctional peptidase 2); proteasome 20S subunit beta 9; proteasome beta 9 subunit; proteasome catalytic subunit 1i; proteasome chain 7; proteasome subunit beta 6i; proteasome subunit beta 9; proteasome subunit beta type 9; proteasome subunit beta type-9; Proteasome subunit beta-1i; proteasome-related gene 2; proteosome (prosome, macropain) subunit, beta type 9; proteosome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2); proteosome beta 9 subunit; PSMB6i; PSMB9; Really interesting new gene 12 protein; Ring12; RING12 protein | |
| Psmb9 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P28076 | |
| 16912 | |
| Recombinant protein Proteasome subunit beta type-9. The antigen corresponds to amino acid range 22-219 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title