missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSMB8 Recombinant Rabbit Monoclonal Antibody (K01_1T51)

Product Code. 30829205
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829205 100 μL
30830403 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829205 Supplier Invitrogen™ Supplier No. MA563530

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DSGYRPNLSP EEAY.

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. This gene is located in the class II region of the MHC (major histocompatibility complex). Expression of this gene is induced by gamma interferon and this gene product replaces catalytic subunit 3 (proteasome beta 5 subunit) in the immunoproteasome. Proteolytic processing is required to generate a mature subunit. Two alternative transcripts encoding two isoforms have been identified; both isoforms are processed to yield the same mature subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB8
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1T51
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene Psmb8
Gene Accession No. P28062, P28063
Gene Alias ALDD; D6S216; D6S216E; DAAP-57C1.3; JMP; large multifunctional protease 7; Lmp7; Lmp-7; low molecular mass polypeptide 7; low molecular mass protein 7; low molecular weight protein 7; macropain subunit C13; Mc13; MC13 (LMP7); MGC1491; multicatalytic endopeptidase complex subunit C13; NKJO; OTTHUMP00000029420; OTTHUMP00000029421; OTTHUMP00000062981; protease component C13; proteasome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteasome (prosome, macropain) subunit, beta type, 8; proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctional peptidase 7); proteasome catalytic subunit 3i; proteasome component C13; proteasome subunit beta 5i; proteasome subunit beta 8; proteasome subunit beta type 8; proteasome subunit beta type-8; Proteasome subunit beta-5i; proteasome subunit MC13; proteasome subunit Y2; proteasome-related gene 7; proteosome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteosome beta 8 subunit; PSMB5i; Psmb8; Really interesting new gene 10 protein; Ring10; Y2
Gene Symbols Psmb8
Host Species Rabbit
Immunogen A synthetic peptide of human LMP7 (aa 212-225).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16913, 5696
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.