missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB7 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-52081
This item is not returnable.
View return policy
Description
PSMB7 Polyclonal specifically detects PSMB7 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Proteasome 20S beta 7 | |
| Polyclonal | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_002790.1 | |
| PSMB7 | |
| Peptide with sequence C-RNAVLEADFAKR corresponding to internal region according to NP_002790.1. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| EC 3.4.25.1, Macropain chain Z, Multicatalytic endopeptidase complex chain Z, proteasome (prosome, macropain) subunit, beta type, 7, proteasome subunit alpha, proteasome subunit beta 7, proteasome subunit beta type-7, Proteasome subunit Z, PSMB7, Zproteasome catalytic subunit 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5695 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction