missing translation for 'onlineSavingsMsg'
Learn More

PSMB5 Antibody, Novus Biologicals™

Product Code. 18327878 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327878 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327878 Supplier Novus Biologicals Supplier No. H00005693D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSMB5 Polyclonal antibody specifically detects PSMB5 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. NP_002788.1
Gene Alias LMPXproteasome beta 5 subunit, Macropain epsilon chain, MB1EC 3.4.25.1, Multicatalytic endopeptidase complex epsilon chain, proteasome (prosome, macropain) subunit, beta type, 5, proteasome catalytic subunit 3, Proteasome chain 6, Proteasome epsilon chain, proteasome subunit beta type-5, Proteasome subunit MB1, Proteasome subunit X, proteasome subunit, beta type, 5, PSX large multifunctional protease X, XMGC104214
Host Species Rabbit
Immunogen PSMB5 (NP_002788.1, 1 a.a. - 263 a.a.) full-length human protein. MALASVLERPLPVNQRGFFGLGGRADLLDLGPGSLSDGLSLAAPGWGVPEEPGIEMLHGTTTLAFKFRHGVIVAADSRATAGAYIASQTVKKVIEINPYLLGTMAGGAADCSFWERLLARQCRIYELRNKERISVAAASKLLANMVYQYKGMGLSMGTMICGWDKRGPGLYYVDSEGNRISGATFSVGSGSVYAYGVMDRGYSYDLEVEQAYDLARRAIYQATYRDAYSGGAVNLYHVREDGWIRVSSDNVADLHEKYSGSTP
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5693
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.