missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13820
This item is not returnable.
View return policy
Description
PSMB5 Polyclonal antibody specifically detects PSMB5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PSMB5 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| LMPXproteasome beta 5 subunit, Macropain epsilon chain, MB1EC 3.4.25.1, Multicatalytic endopeptidase complex epsilon chain, proteasome (prosome, macropain) subunit, beta type, 5, proteasome catalytic subunit 3, Proteasome chain 6, Proteasome epsilon chain, proteasome subunit beta type-5, Proteasome subunit MB1, Proteasome subunit X, proteasome subunit, beta type, 5, PSX large multifunctional protease X, XMGC104214 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: YRDAYSGGAVNLYHVREDGWIRVSSDNVADLHEKYSGST | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 5693 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction