missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35905-20ul
This item is not returnable.
View return policy
Description
PSMB5 Polyclonal antibody specifically detects PSMB5 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| PSMB5 | |
| Polyclonal | |
| Western Blot 1:2000 - 1:7000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:50 - 1:100, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| LMPXproteasome beta 5 subunit, Macropain epsilon chain, MB1EC 3.4.25.1, Multicatalytic endopeptidase complex epsilon chain, proteasome (prosome, macropain) subunit, beta type, 5, proteasome catalytic subunit 3, Proteasome chain 6, Proteasome epsilon chain, proteasome subunit beta type-5, Proteasome subunit MB1, Proteasome subunit X, proteasome subunit, beta type, 5, PSX large multifunctional protease X, XMGC104214 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-263 of human PSMB5 (NP_002788.1).,, Sequence:, MALASVLERPLPVNQRGFFGLGGRADLLDLGPGSLSDGLSLAAPGWGVPEEPGIEMLHGTTTLAFKFRHGVIVAADSRATAGAYIASQTVKKVIEINPYLLGTMAGGAADCSFWERLLARQCRIYELRNKERISVAAASKLLANMVYQYKGMGLSMGTMICGWDKRGPGLYYVDSEGNRISGATFSVGSGSVYAYGVMDRGYSYDLEVEQAYDLARRAIYQATYRDAYSGGAVNLYHVREDGWIRVSSDNVADLHEKYSGSTP | |
| 20 μL | |
| Core ESC Like Genes, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway | |
| 5693 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction