Learn More
Invitrogen™ PSMB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528999
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (90%), Xenopus laevis (91%), Dog (99%), Pig (97%), Rhesus Monkey (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. Pseudogenes have been identified on chromosomes 2 and 12.
Specifications
| PSMB3 | |
| Polyclonal | |
| Unconjugated | |
| PSMB3 | |
| AL033320; C10-II; HC10-II; MGC4147; proteasome (prosome, macropain) subunit, beta type 3; proteasome (prosome, macropain) subunit, beta type, 3; proteasome beta 3 subunit; proteasome chain 13; Proteasome component C10-II; proteasome subunit beta 3; proteasome subunit beta type-3; Proteasome theta chain; PSMB3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5691 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.96 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P49720 | |
| PSMB3 | |
| Full length human Proteasome 20S beta 3 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.