missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555305
This item is not returnable.
View return policy
Description
Immunogen sequence: YVLVASDRVA ASNIVQMKDD HDKMFKMSEK ILLLCVGEAG DTVQFAEYIQ KNVQLYKMRN GYELSPTAAA NFTRRNLADC LRSRTP Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity. This subunit has a trypsin-like activity.
Specifications
| PSMB2 | |
| Polyclonal | |
| Unconjugated | |
| PSMB2 | |
| AU045357; AW108089; C7-I; D4Wsu33e; HC7-I; Macropain subunit C7-I; multicatalytic endopeptidase complex subunit C7-1; multicatalytic endopeptidase complex subunit C7-I; proteasome (prosome, macropain) subunit, beta type 2; proteasome (prosome, macropain) subunit, beta type, 2; proteasome beta 2 subunit; proteasome component C7-I; proteasome subunit beta 2; proteasome subunit beta type-2; proteasome subunit, beta type, 2; PSMB2; testicular tissue protein Li 152 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5690 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49721 | |
| PSMB2 | |
| Recombinant protein corresponding to Human PSMB2. Recombinant protein control fragment (Product #RP-94169). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction