missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB10 Recombinant Rabbit Monoclonal Antibody (23GB2170), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA552481
This item is not returnable.
View return policy
Description
PSMB10 Recombinant Monoclonal antibody specifically detects PSMB10 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
| PSMB10 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35955, P40306, Q4KM35 | |
| PSMB10 | |
| A synthesized peptide derived from human PSMB10 (215-250AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2170 | |
| Unconjugated | |
| PSMB10 | |
| beta2i; LMP10; Low molecular mass protein 10; macropain; Macropain subunit MECl-1; MECL1; Mecl-1; MGC1665; multicatalytic endopeptidase complex subunit MECl-1; OTTHUMP00000174858; prosome; prosome Mecl1; proteasome (prosome, macropain) subunit, beta type 10; proteasome (prosome, macropain) subunit, beta type, 10; proteasome (prosomome, macropain) subunit, beta type 10; proteasome 20S subunit beta 10; proteasome beta 10 subunit; proteasome catalytic subunit 2i; Proteasome MECl 1; proteasome MECl-1; proteasome subunit 10; proteasome subunit beta 10; proteasome subunit beta 7i; proteasome subunit beta type-10; Proteasome subunit beta-2i; proteasome subunit LMP10; proteasome subunit MECL1; PSMB 10; Psmb10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19171, 291983, 5699 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction