missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMB10 Recombinant Rabbit Monoclonal Antibody (23GB2165), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | PSMB10 |
|---|---|
| Clone | 23GB2165 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089196
![]() |
Invitrogen™
MA553238 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089850
|
Invitrogen™
MA553239 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSMB10 Recombinant Monoclonal antibody specifically detects PSMB10 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| PSMB10 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O35955, P40306, Q4KM35 | |
| 19171, 291983, 5699 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2165 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| beta2i; LMP10; Low molecular mass protein 10; macropain; Macropain subunit MECl-1; MECL1; Mecl-1; MGC1665; multicatalytic endopeptidase complex subunit MECl-1; OTTHUMP00000174858; prosome; prosome Mecl1; proteasome (prosome, macropain) subunit, beta type 10; proteasome (prosome, macropain) subunit, beta type, 10; proteasome (prosomome, macropain) subunit, beta type 10; proteasome 20S subunit beta 10; proteasome beta 10 subunit; proteasome catalytic subunit 2i; Proteasome MECl 1; proteasome MECl-1; proteasome subunit 10; proteasome subunit beta 10; proteasome subunit beta 7i; proteasome subunit beta type-10; Proteasome subunit beta-2i; proteasome subunit LMP10; proteasome subunit MECL1; PSMB 10; Psmb10 | |
| A synthesized peptide derived from human PSMB10 (50-80AA). | |
| Primary | |
| -20°C | |
| PSMB10 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title