missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB10/MECL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £411.00
Specifications
| Antigen | PSMB10/MECL1 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:2500-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18433391
|
Novus Biologicals
NBP1-88660-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18790524
|
Novus Biologicals
NBP1-88660 |
0.1 mL |
£411.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSMB10/MECL1 Polyclonal specifically detects PSMB10/MECL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PSMB10/MECL1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| beta2i, FLJ00366, LMP10Proteasome subunit beta-2i, Low molecular mass protein 10, Macropain subunit MECl-1, MECL1EC 3.4.25.1, MGC1665, Multicatalytic endopeptidase complex subunit MECl-1, proteasome (prosome, macropain) subunit, beta type, 10, proteasome catalytic subunit 2i, Proteasome MECl-1, proteasome subunit beta 7i, proteasome subunit beta type-10, proteasome subunit MECL1 | |
| PSMB10 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:2500-1:5000 | |
| Polyclonal | |
| Rabbit | |
| Immunology | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5699 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AGILGDLGSGGNVDACVITKTGAKLLRTLSSPTEPVKRSGRYHFVPGTTAVLTQTVKPLTLELVEETVQAMEVE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title