missing translation for 'onlineSavingsMsg'
Learn More

PSMB10/MECL1 Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30730894 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30730894 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30730894 Supplier Novus Biologicals Supplier No. AF7684JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

PSMB10/MECL1 Polyclonal antibody specifically detects PSMB10/MECL1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB10/MECL1
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias beta2i, FLJ00366, LMP10Proteasome subunit beta-2i, Low molecular mass protein 10, Macropain subunit MECl-1, MECL1EC 3.4.25.1, MGC1665, Multicatalytic endopeptidase complex subunit MECl-1, proteasome (prosome, macropain) subunit, beta type, 10, proteasome catalytic subunit 2i, Proteasome MECl-1, proteasome subunit beta 7i, proteasome subunit beta type-10, proteasome subunit MECL1
Host Species Goat
Immunogen E. coli-derived recombinant human PSMB10/MECL1, Thr40-Glu273, Accession # P40306
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5699
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.