missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102335
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PSMB1.
PSMB1 is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity.
Specifications
| PSMB1 | |
| Polyclonal | |
| Unconjugated | |
| PSMB1 | |
| AA409053; C81484; component C5 of proteasome; HC5; Lmpc5; Macropain subunit C5; multicatalytic endopeptidase complex subunit C5; PMSB1; proteasome (prosome, macropain) subunit, beta type 1; proteasome (prosome, macropain) subunit, beta type, 1; proteasome beta 1 subunit; proteasome beta-subunit C5; proteasome component C5; Proteasome gamma chain; proteasome subunit beta 1; proteasome subunit beta type-1; proteasome subunit C5; proteasome subunit HC5; proteasome subunit RC5; PSC5; Psmb1; testicular secretory protein Li 45 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19170, 5689, 94198 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O09061, P18421, P20618 | |
| PSMB1 | |
| A synthesized peptide derived from human PSMB1(Accession P20618), corresponding to amino acid residues Y135-N185. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction