missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMAL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524990
This item is not returnable.
View return policy
Description
PSMAL Polyclonal Antibody for Western Blot, IHC (P), Flow
PSMAL has both folate hydrolase and N-acetylated-alpha-linked-acidic dipeptidase (NAALADase) activity. PSMAL exhibits a dipeptidyl-peptidase IV type activity.
Specifications
| PSMAL | |
| Polyclonal | |
| Unconjugated | |
| FOLH1B | |
| cell growth-inhibiting gene 26 protein; folate hydrolase 1B; folate hydrolase 2; folate hydrolase pseudogene; FOLH1B; FOLH2; FOLHP; GCP3; GCPIII; GIG26; glutamate carboxypeptidase III; NAALADase; N-acetylated-alpha-linked-acidic dipeptidase; Prostate-specific membrane antigen-like protein; PSM; PSMAL; PSMA-LIKE; Putative folate hydrolase 1B; putative N-acetylated-alpha-linked acidic dipeptidase | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 219595 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9HBA9 | |
| FOLH1B | |
| KLH conjugated synthetic peptide between 155-183 amino acids from the central region of human PSMAL. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction