missing translation for 'onlineSavingsMsg'
Learn More

PSMA8 Antibody, Novus Biologicals™

Product Code. 18334850 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334850 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334850 Supplier Novus Biologicals Supplier No. NBP286758

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSMA8 Polyclonal specifically detects PSMA8 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen PSMA8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias EC 3.4.25.1, MGC26605, proteasome (prosome, macropain) subunit, alpha type, 8, proteasome subunit alpha type-7-like, PSMA7LMGC26605
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of Human PSMA8
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143471
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.