missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMA4 Monoclonal Antibody (OTI4B8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525812
This item is not returnable.
View return policy
Description
PSMA4 Monoclonal Antibody for Western Blot, IHC (P)
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Three alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| PSMA4 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21670, P25789, Q9R1P0 | |
| Psma4 | |
| Full length human recombinant protein of PSMA4 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B8 | |
| Unconjugated | |
| Psma4 | |
| C9; HC9; HsT17706; LOW QUALITY PROTEIN: proteasome subunit alpha type-4; macropain subunit C9; MGC111191; MGC12467; MGC24813; Multicatalytic endopeptidase complex subunit C9; multicatalytic proteinase subunit L; proteasome (prosome, macropain) subunit, alpha type 4; proteasome (prosome, macropain) subunit, alpha type, 4; proteasome 20S subunit alpha 4; proteasome alpha 4 subunit; proteasome component C9; proteasome subunit alpha 4; proteasome subunit alpha type-4; proteasome subunit alpha type-4-like protein; proteasome subunit HC9; proteasome subunit L; PSC9; PSMA 4; PSMA4; QtsA-10816; unnamed protein product; wu:fe05d10; zgc:56176 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26441, 29671, 5685 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction