missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMA3 Recombinant Rabbit Monoclonal Antibody (17H20L21)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 703791
This item is not returnable.
View return policy
Description
This antibody is predicted to react with Rat, Bovine.
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Two alternative transcripts encoding different isoforms have been identified.
Specifications
| PSMA3 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O70435, P25788 | |
| Psma3 | |
| Peptide corresponding to human PSMA3 [1) aa27-aa41 2) aa220-aa235]. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 17H20L21 | |
| Unconjugated | |
| Psma3 | |
| HC8; Lmpc8; Macropain subunit C8; multicatalytic endopeptidase complex subunit C8; multicatalytic proteinase subunit K; proteasome (prosome, macropain) subunit, alpha type 3; proteasome (prosome, macropain) subunit, alpha type, 3; proteasome component C8; proteasome subunit alpha 3; proteasome subunit alpha type-3; proteasome subunit C8; proteasome subunit K; PSA3; PSC3; PSC8; PSMA3; Psma3l; testicular secretory protein Li 43 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 19167, 5684 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction