Learn More
PSMA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11887-1-AP-150UL
 View more versions of this product
Product Code. 16869253
  • £349.00 / 150µL
Description
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Two alternative transcripts encoding different isoforms have been identified.
Specifications
Specifications
PSMA3
Polyclonal
Unconjugated
PSMA3
20S proteasome, HC8, Macropain subunit C8, Proteasome component C8, PSC3, PSC8, PSMA3
Rabbit
Antigen Affinity Chromatography
RUO
19167, 29670, 5684
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O70435, P18422, P25788
Psma3
PSMA3 Fusion Protein Ag2495
150 μL
Primary
Human, Mouse, Rat