missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579890
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat testis tissue, mouse lung tissue, 293T whole cell. IHC: mouse brain tissue, rat brain tissue, human intestinal cancer tissue. Flow: HeLa cell.
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Two alternative transcripts encoding different isoforms have been identified.
Specifications
| PSMA3 | |
| Polyclonal | |
| Unconjugated | |
| Psma3 | |
| HC8; Lmpc8; Macropain subunit C8; multicatalytic endopeptidase complex subunit C8; multicatalytic proteinase subunit K; proteasome (prosome, macropain) subunit, alpha type 3; proteasome (prosome, macropain) subunit, alpha type, 3; proteasome component C8; proteasome subunit alpha 3; proteasome subunit alpha type-3; proteasome subunit C8; proteasome subunit K; PSA3; PSC3; PSC8; PSMA3; Psma3l; testicular secretory protein Li 43 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19167, 29670, 5684 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| O70435, P18422, P25788 | |
| Psma3 | |
| A synthetic peptide corresponding to a sequence in the middle region of human PSMA3 (88-127aa LADIAREEASNFRSNFGYNIPLKHLADRVAMYVHAYTLYS). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction