missing translation for 'onlineSavingsMsg'
Learn More

PSMA2 Antibody [Biotin], Novus Biologicals™

Product Code. 30735647 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735647 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735647 Supplier Novus Biologicals Supplier No. AF7639B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

PSMA2 Polyclonal antibody specifically detects PSMA2 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMA2
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias HC3;MU;PMSA2;Proteasome subunit alpha type;Proteasome subunit alpha type-2;PSC2
Host Species Sheep
Immunogen E. coli-derived recombinant human PSMA2, Ala2-Ala234, Accession # P25787
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5683
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.