Learn More
PSMA1 Mouse anti-Human, Unconjugated, Clone: 1D1D5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67910-1-IG-20UL
Description
The PSMA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PSMA1. This antibody recognizes human, mouse, rat antigen. The PSMA1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PSMA1 | |
| Monoclonal | |
| Unconjugated | |
| 20 μL | |
| 5682 | |
| IgG2b |
| Western Blot, Immunohistochemistry, ELISA | |
| 1D1D5 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |