missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-624
This item is not returnable.
View return policy
Description
PSMA1 Polyclonal specifically detects PSMA1 in Human, Mouse, Rat, Canine, Chicken, Chinese Hamster samples. It is validated for Western Blot.
Specifications
| Proteasome 20S C2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| PSMA1 | |
| Synthetic Peptide: C P(249) A D E P A E K A D E P M E H(263) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects proteasome 20S C2 subunit. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 ug/ml | |
| EC 3.4.25.1, HC2Proteasome nu chain, Macropain subunit C2, macropain subunit nu, MGC14542, MGC14575, MGC14751, MGC1667, MGC21459, MGC22853, MGC23915, Multicatalytic endopeptidase complex subunit C2, NU30 kDa prosomal protein, PROS-30, PROS30Proteasome component C2, proteasome (prosome, macropain) subunit, alpha type, 1, proteasome subunit alpha type-1, proteasome subunit nu, proteasome subunit, alpha-type, 1, protein P30-33K, PSC2 | |
| Rabbit | |
| 30 kDa | |
| 100 μg | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5682 | |
| Human, Mouse, Rat, Canine, Chicken, Chinese Hamster | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction