missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109222
This item is not returnable.
View return policy
Description
Positive Control: Human small intestine tissue lysate, HepG2, PC-3M.
This gene encodes a type II transmembrane glycoprotein belonging to the M28 peptidase family. The protein acts as a glutamate carboxypeptidase on different alternative substrates, including the nutrient folate and the neuropeptide N-acetyl-l-aspartyl-l-glutamate and is expressed in a number of tissues such as prostate, central and peripheral nervous system and kidney. A mutation in this gene may be associated with impaired intestinal absorption of dietary folates, resulting in low blood folate levels and consequent hyperhomocysteinemia. Expression of this protein in the brain may be involved in a number of pathological conditions associated with glutamate excitotoxicity. In the prostate the protein is up-regulated in cancerous cells and is used as an effective diagnostic and prognostic indicator of prostate cancer. This gene likely arose from a duplication event of a nearby chromosomal region. Alternative splicing gives rise to multiple transcript variants encoding several different isoforms.
Specifications
| PSMA | |
| Polyclonal | |
| Unconjugated | |
| FOLH1 | |
| Cell growth-inhibiting gene 27 protein; FGCP; folate hydrolase (prostate-specific membrane antigen) 1; folate hydrolase 1; FOLH; Folh1; Folylpoly-gamma-glutamate carboxypeptidase; GCP2; GCPII; GIG27; glutamate carboxylase II; glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; membrane glutamate carboxypeptidase; mGCP; mopsm; Naalad; NAALAD1; NAALAdase; NAALADase I; N-acetylated alpha-linked acidic dipeptidase; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; prostate specific membrane antigen; prostate specific membrane antigen variant F; prostate-specific membrane antigen; Prostate-specific membrane antigen homolog; PSM; PSMA; pteroylpoly-gamma-glutamate carboxypeptidase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2346 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q04609 | |
| FOLH1 | |
| Synthetic peptide within Human PSMA aa 676-725. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction