missing translation for 'onlineSavingsMsg'
Learn More

PSMA/FOLH1/NAALADase I Antibody (FOLH1/2354), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08385R

Product Code. 18593244

  • £462.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PSMA/FOLH1/NAALADase I Monoclonal antibody specifically detects PSMA/FOLH1/NAALADase I in Human, Canine, Equine, Feline samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PSMA/FOLH1/NAALADase I
Monoclonal
DyLight 550
Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase
Recombinant human PSMA/FOLH1/NAALADase I protein fragment (around aa 232-433) (exact sequence is proprietary) (Uniprot: Q04609 )
100 μg
Cancer, Prostate Cancer
2346
Store at 4°C in the dark.
IgG2b κ
Western Blot, Immunohistochemistry (Paraffin), Protein Array
FOLH1/2354
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Canine, Equine, Feline
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.