Learn More
Invitrogen™ PSKH1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA599695
Product Code. 16613945
  • £448.00 / 100µL
Description
Antibody detects endogenous levels of total PSK-H1.
PSKH1 may be a SFC-associated serine kinase (splicing factor compartment-associated serine kinase) with a role in intranuclear SR protein (non-snRNP splicing factors containing a serine/arginine-rich domain) trafficking and pre-mRNA processing. PSKH1 is localized in the Brefeldin A-sensitive Golgi compartment, at centrosomes, in the nucleus with a somewhat speckle-like presence, membrane-associated to the endoplasmic reticulum (ER) and the plasma membrane (PM), and more diffusely in the cytoplasm.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PSKH1
Polyclonal
Unconjugated
Pskh1
AW539964; b2b1230Clo; E130013P03Rik; Mutant line 1230; protein serine kinase H1; PSKH1; PSK-H1; Serine/threonine-protein kinase H1
Rabbit
Affinity chromatography
RUO
364993, 5681
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P11801
Pskh1
A synthesized peptide derived from human PSKH1(Accession P11801), corresponding to amino acid residues T275-S325.
100 μL
Primary
Human, Rat