Learn More
Invitrogen™ PSKH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585311
Description
Keep as concentrated solution. Predicted reactivity: Mouse (96%), Xenopus laevis (80%), Chicken (81%), Rhesus Monkey (99%), Bovine (95%). Positive Control: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PSKH1 may be a SFC-associated serine kinase (splicing factor compartment-associated serine kinase) with a role in intranuclear SR protein (non-snRNP splicing factors containing a serine/arginine-rich domain) trafficking and pre-mRNA processing. PSKH1 is localized in the Brefeldin A-sensitive Golgi compartment, at centrosomes, in the nucleus with a somewhat speckle-like presence, membrane-associated to the endoplasmic reticulum (ER) and the plasma membrane (PM), and more diffusely in the cytoplasm.
Specifications
| PSKH1 | |
| Polyclonal | |
| Unconjugated | |
| Pskh1 | |
| AW539964; b2b1230Clo; E130013P03Rik; Mutant line 1230; protein serine kinase H1; PSKH1; PSK-H1; Serine/threonine-protein kinase H1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 244631, 5681 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P11801, Q91YA2 | |
| Pskh1 | |
| Recombinant protein encompassing a sequence within the center region of human PSKH1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.