missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Content And Storage | Storage is content dependent. |
| Quantity | 25 Tests |
| Conjugate | IANBD |
| Kit Contents | pSIVA-IANBD, Propidium Iodide Staining Solution, 10X PBS Buffer, 10X Binding Buffer |
| For Use With (Application) | Flow Cytometry, Flow (Cell Surface) |
| Product Type | Flow Assay Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?