missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ pSIVA Apoptosis Detection Flow Cytometry Kit

Product Code. 18031374 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18031374 100 Tests 100 tests
18181763 25 Tests 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only
Product Code. 18031374 Supplier Novus Biologicals™ Supplier No. NBP229611100Tests

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only

Applications: Flow Cytometry, Flow (Cell Surface)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage is content dependent.
Quantity 100 Tests
Conjugate IANBD
Kit Contents pSIVA-IANBD, Propidium Iodide Staining Solution, 10X PBS Buffer, 10X Binding Buffer
For Use With (Application) Flow Cytometry, Flow (Cell Surface)
Product Type Flow Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.