missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSIP1 Recombinant Rabbit Monoclonal Antibody (24GB840)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557648
This item is not returnable.
View return policy
Description
PSIP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Transcriptional coactivator involved in neuroepithelial stem cell differentiation and neurogenesis. Involved in particular in lens epithelial cell gene regulation and stress responses. May play an important role in lens epithelial to fiber cell terminal differentiation. May play a protective role during stress-induced apoptosis. Isoform 2 is a more general and stronger transcriptional coactivator. Isoform 2 may also act as an adapter to coordinate pre-mRNA splicing. Cellular cofactor for lentiviral integration.
Specifications
| PSIP1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PSIP1 | |
| AA408851; AU015605; CLL-associated antigen KW-7; dense fine speckles 70; dense fine speckles 70 kDa protein; DFS 70; DFS70; LEDGF; Ledgfa; Ledgfb; Lens epithelium-derived growth factor; mLEDGF; p52; p75; PAIP; PC4 and SFRS1 interacting protein 1; PC4 and SFRS1 interacting protein 2; PC4 and SFRS1-interacting protein; PSIP1; Psip2; SBP75; supercoiled DNA binding protein 75; transcriptional coactivator p52/p75; Transcriptional coactivator p75/p52 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 101739, 11168, 313323 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB840 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75475, Q812D1, Q99JF8 | |
| PSIP1 | |
| A synthesized peptide derived from human PSIP1 / LEDGF (aa 1-61). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction