missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554060
This item is not returnable.
View return policy
Description
Immunogen sequence: KQVETEEAGV VTTATASVNL KVSPKRGRPA ATEVKIPKPR GRPKMVKQPC PSESDIITEE DKSKKKGQEE KQPKKQPKKD EEGQ Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 83%.
Transcriptional coactivator involved in neuroepithelial stem cell differentiation and neurogenesis. Involved in particular in lens epithelial cell gene regulation and stress responses. May play an important role in lens epithelial to fiber cell terminal differentiation. May play a protective role during stress-induced apoptosis. Isoform 2 is a more general and stronger transcriptional coactivator. Isoform 2 may also act as an adapter to coordinate pre-mRNA splicing. Cellular cofactor for lentiviral integration.
Specifications
| PSIP1 | |
| Polyclonal | |
| Unconjugated | |
| PSIP1 | |
| AA408851; AU015605; CLL-associated antigen KW-7; dense fine speckles 70; dense fine speckles 70 kDa protein; DFS 70; DFS70; LEDGF; Ledgfa; Ledgfb; Lens epithelium-derived growth factor; mLEDGF; p52; p75; PAIP; PC4 and SFRS1 interacting protein 1; PC4 and SFRS1 interacting protein 2; PC4 and SFRS1-interacting protein; PSIP1; Psip2; SBP75; supercoiled DNA binding protein 75; transcriptional coactivator p52/p75; Transcriptional coactivator p75/p52 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11168 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75475 | |
| PSIP1 | |
| Recombinant protein corresponding to Human PSIP1. Recombinant protein control fragment (Product #RP-92573). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction