missing translation for 'onlineSavingsMsg'
Learn More

PSI-7409 tetrasodium, MedChemExpress

Product Code. 16496788
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496788 10 mg 10mg
16493486 25 mg 25mg
16479337 5 mg 5mg
16461549 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496788 Supplier Medchem Express Supplier No. HY15745A
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PSI-7409 tetrasodium is an active 5'-triphosphate metabolite of sofosbuvir (PSI-7977), inhibiting HCV NS5B polymerases, with IC50s of 1.6, 2.8, 0.7 and 2.6 μM for GT 1b_Con1, GT 2a_JFH1, GT 3a, and GT 4a NS5B polymerases, respectively.

PSI-7409 tetrasodium is an active 5'-triphosphate metabolite of sofosbuvir (PSI-7977), inhibiting HCV NS5B polymerases, with IC50s of 1.6, 2.8, 0.7 and 2.6 μM for GT 1b_Con1, GT 2a_JFH1, GT 3a, and GT 4a NS5B polymerases, respectively.

Specifications

Chemical Name or Material PSI-7409 tetrasodium
CAS 1621884-22-7
Color White
Purity Grade Notes Research
Molecular Formula C10H12FN2Na4O14P3
Quantity 10 mg
Solubility Information H2O : ≥ 125 mg/mL (212.55 mM)
SMILES O[C@@H]([C@@](C)(F)[C@H](N1C(NC(C=C1)=O)=O)O2)[C@H]2COP(OP(OP(O[Na])(O[Na])=O)(O[Na])=O)(O[Na])=O
Molecular Weight (g/mol) 588.09
Formula Weight 588.09
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.