missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSI-7409 tetrasodium, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
Description
PSI-7409 tetrasodium is an active 5'-triphosphate metabolite of sofosbuvir (PSI-7977), inhibiting HCV NS5B polymerases, with IC50s of 1.6, 2.8, 0.7 and 2.6 μM for GT 1b_Con1, GT 2a_JFH1, GT 3a, and GT 4a NS5B polymerases, respectively.
Specifications
Specifications
| Chemical Name or Material | PSI-7409 tetrasodium |
| CAS | 1621884-22-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C10H12FN2Na4O14P3 |
| Quantity | 5 mg |
| Solubility Information | H2O : ≥ 125 mg/mL (212.55 mM) |
| SMILES | O[C@@H]([C@@](C)(F)[C@H](N1C(NC(C=C1)=O)=O)O2)[C@H]2COP(OP(OP(O[Na])(O[Na])=O)(O[Na])=O)(O[Na])=O |
| Molecular Weight (g/mol) | 588.09 |
| Formula Weight | 588.09 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction